missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106935
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DAP.
This gene encodes a basic, proline-rich, 15-kD protein. The protein acts as a positive mediator of programmed cell death that is induced by interferon-gamma.
Specifications
| DAP | |
| Polyclonal | |
| Unconjugated | |
| DAP | |
| 4921531N22Rik; AI789753; amylin peptide; AU040360; DAP; DAP 1; DAP1; DAP-1; death associated protein; death-associated protein; death-associated protein 1; MGC99796; OTTHUMP00000161670; OTTHUMP00000221331; Rap7a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1611, 223453, 64322 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51397, Q91XC8, Q9QX67 | |
| DAP | |
| A synthesized peptide derived from human DAP(Accession P51397), corresponding to amino acid residues M1-K37. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction