missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP10/HCST Antibody (982107), DyLight 755, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB97862IR
This item is not returnable.
View return policy
Description
DAP10/HCST Monoclonal specifically detects DAP10/HCST in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| DAP10/HCST | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HCST | |
| Synthetic peptide containing human DAP10/HCST | |
| 0.1 mL | |
| Adaptive Immunity, Immune System Diseases, Immunology, Innate Immunity | |
| Human | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 982107 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DAP10PIK3AP, DKFZP586C1522, hematopoietic cell signal transducer, KAP10DNAX-activation protein 10, kinase assoc pro of ∽10kDa, kinase assoc protein, Membrane protein DAP10, phosphoinositide-3-kinase adaptor protein, Transmembrane adapter protein KAP10 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction