missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP12 Mouse anti-Human, Alexa Fluor™ 647, Clone: 406288, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566603
This item is not returnable.
View return policy
Description
DAP12 Monoclonal antibody specifically detects DAP12 in Human samples. It is validated for Flow Cytometry.
Specifications
| DAP12 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| DAP12 | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 406288 | |
| Alexa Fluor 647 | |
| 25 μg | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction