missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP3 Antibody (42C617.1.2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56539H
This item is not returnable.
View return policy
Description
DAP3 Monoclonal specifically detects DAP3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| DAP3 | |
| Monoclonal | |
| HRP | |
| P51398 | |
| DAP3 | |
| The antibody was raised against a synthetic peptide corresponding to amino acids 51-66 of human DAP-3 protein. | |
| 0.1 mL | |
| Apoptosis, Core ESC Like Genes, Stem Cell Markers | |
| 7818 | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 42C617.1.2 | |
| Western Blot | |
| bMRP-10, DAP-3Death-associated protein 3, death associated protein 3, DKFZp686G12159,28S ribosomal protein S29, mitochondrial, FLJ12817, MGC126058, MGC126059, mitochondrial 28S ribosomal protein S29, MRP-S29Ionizing radiation resistance conferring protein, MRPS29S29mt | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In Jurkat, a 45 kDa band is observed. | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction