missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP3 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98784R
This item is not returnable.
View return policy
Description
DAP3 Polyclonal specifically detects DAP3 in Human samples. It is validated for Western Blot.
Specifications
| DAP3 | |
| Polyclonal | |
| Western Blot | |
| bMRP-10, DAP-3Death-associated protein 3, death associated protein 3, DKFZp686G12159,28S ribosomal protein S29, mitochondrial, FLJ12817, MGC126058, MGC126059, mitochondrial 28S ribosomal protein S29, MRP-S29Ionizing radiation resistance conferring protein, MRPS29S29mt | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human DAP3. | |
| 0.1 mL | |
| Apoptosis, Core ESC Like Genes, Stem Cell Markers | |
| 7818 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction