missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAP3 Antibody (OTI1B12), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71338FR
This item is not returnable.
View return policy
Description
DAP3 Monoclonal specifically detects DAP3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| DAP3 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| DAP3 | |
| Recombinant protein fragment corresponding to amino acids 161-398 of human DAP3 (NP_387506) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1B12 | |
| Western Blot, Immunohistochemistry | |
| bMRP-10, DAP-3Death-associated protein 3, death associated protein 3, DKFZp686G12159,28S ribosomal protein S29, mitochondrial, FLJ12817, MGC126058, MGC126059, mitochondrial 28S ribosomal protein S29, MRP-S29Ionizing radiation resistance conferring protein, MRPS29S29mt | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Core ESC Like Genes, Stem Cell Markers | |
| 7818.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction