Learn More
DAP3 Monoclonal Antibody (OTI2D4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802818S
Encompass_Preferred
 View more versions of this product
Product Code. 17541563
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
DAP3 Monoclonal antibody specifically detects DAP3 in Human samples. It is validated for Western Blot
Specifications
Specifications
DAP3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NY93
Mouse
Affinity chromatography
RUO
54606
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI2D4
Unconjugated
DDX56
28S ribosomal protein S29, mitochondrial; 4921514D13Rik; bMRP-10; DAP3; DAP-3; death associated protein 3; death-associated protein 3; DKFZp686G12159; Ionizing radiation resistance conferring protein; MGC126058; MGC126059; mitochondrial 28S ribosomal protein S29; Mitochondrial small ribosomal subunit protein mS29; MRPS29; MRP-S29; S29mt
Human recombinant protein fragment corresponding to amino acids 323-547 of human DDX56 produced in E.coli.
30 μL