missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dapagliflozin Tetraacetate ([(2R,3R,4R,5S,6S)-3,4,5-Triacetoxy-6-[4-chloro-3-(4-ethoxybenzyl)phenyl]tetrahydropyran-2-yl]methyl Acetate), Mikromol™
Description
Dapagliflozin Tetraacetate ([(2R,3R,4R,5S,6S)-3,4,5-Triacetoxy-6-[4-chloro-3-(4-ethoxybenzyl)phenyl]tetrahydropyran-2-yl]methyl Acetate)
Specifications
Specifications
| Chemical Name or Material | Dapagliflozin Tetraacetate |
| CAS | 461432-25-7 |
| Analyte or Component Names | Dapagliflozin Tetraacetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H33 Cl O10 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Hydromorphone N-Oxide Hydrochloride, ([(2R,3R,4R,5S,6(S)-3,4,5-Triacetoxy-6-[4-chloro-3-(4-ethoxybenzyl)phenyl]tetrahydropyran-2-yl]methyl Acetate), (1S)-1,5-Anhydro-1-C-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-D-glucitol 2,3,4,6-tetraacetate, D-Glucitol, 1,5-anhydro-1-C-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-, tetraacetate, (1S)- (9CI) |
| SMILES | CCOc1ccc(Cc2cc(ccc2Cl)[C@@H]3O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]3OC(=O)C)cc1 |
| Molecular Weight (g/mol) | 577.02 |
| Quantity | 25 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?