missing translation for 'onlineSavingsMsg'
Learn More

Dapiprazole hydrochloride, MedChemExpress

Product Code. 16484187
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16484187

Brand: Medchem Express HYA0142A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dapiprazole hydrochloride is a potent, selective and orally active alpha-1 adrenoceptor antagonist. Dapiprazole hydrochloride suppresses the opioid withdrawal symptoms. Dapiprazole hydrochloride is also used as eye drops for reversing mydriasis.

Dapiprazole hydrochloride is a potent, selective and orally active alpha-1 adrenoceptor antagonist. Dapiprazole hydrochloride suppresses the opioid withdrawal symptoms. Dapiprazole hydrochloride is also used as eye drops for reversing mydriasis.

Specifications

Chemical Name or Material Dapiprazole hydrochloride
Physical Form Solid
CAS 72822-13-0
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H28ClN5
Solubility Information H2O : 50 mg/mL (138.16 mM; Need ultrasonic) ∣DMSO : ≥ 31 mg/mL (85.66 mM)
SMILES CC1=CC=CC=C1N2CCN(CCC3=NN=C4CCCCN43)CC2.[H]Cl
Molecular Weight (g/mol) 361.91
Formula Weight 361.91
Percent Purity 99.44%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.