missing translation for 'onlineSavingsMsg'
Learn More

Dapiprazole Hydrochloride, TRC

Product Code. 30356290
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30356290

Brand: Toronto Research Chemicals TRCD18545050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dapiprazole Hydrochloride
CAS 72822-13-0
Molecular Formula C19 H27 N5 . Cl H
Quantity 50 mg
Synonym 1,2,4-Triazolo[4,3-a]pyridine, 5,6,7,8-tetrahydro-3-[2-[4-(2-methylphenyl)-1-piperazinyl]ethyl]-, hydrochloride (1:1), 1,2,4-Triazolo[4,3-a]pyridine, 5,6,7,8-tetrahydro-3-[2-[4-(2-methylphenyl)-1-piperazinyl]ethyl]-, monohydrochloride (9CI), AF 2139, Glamidolo, Rev-Eyes, Reversil, 5,6,7,8-Tetrahydro-3-[2-[4-(2-methylphenyl)-1-piperazinyl]ethyl]-1,2,4-Triazolo[4,3-a]pyridine Hydrochloride
InChI Formula InChI=1S/C19H27N5.ClH/c1-16-6-2-3-7-17(16)23-14-12-22(13-15-23)11-9-19-21-20-18-8-4-5-10-24(18)19;/h2-3,6-7H,4-5,8-15H2,1H3;1H
SMILES Cl.Cc1ccccc1N2CCN(CCc3nnc4CCCCn34)CC2
IUPAC Name 3-[2-[4-(2-methylphenyl)piperazin-1-yl]ethyl]-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyridine;hydrochloride
Molecular Weight (g/mol) 361.9121
Formula Weight 361.2033
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.