missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dapiprazole Hydrochloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Dapiprazole Hydrochloride |
| CAS | 72822-13-0 |
| Molecular Formula | C19 H27 N5 . Cl H |
| Quantity | 500 mg |
| Synonym | 1,2,4-Triazolo[4,3-a]pyridine, 5,6,7,8-tetrahydro-3-[2-[4-(2-methylphenyl)-1-piperazinyl]ethyl]-, hydrochloride (1:1), 1,2,4-Triazolo[4,3-a]pyridine, 5,6,7,8-tetrahydro-3-[2-[4-(2-methylphenyl)-1-piperazinyl]ethyl]-, monohydrochloride (9CI), AF 2139, Glamidolo, Rev-Eyes, Reversil, 5,6,7,8-Tetrahydro-3-[2-[4-(2-methylphenyl)-1-piperazinyl]ethyl]-1,2,4-Triazolo[4,3-a]pyridine Hydrochloride |
| InChI Formula | InChI=1S/C19H27N5.ClH/c1-16-6-2-3-7-17(16)23-14-12-22(13-15-23)11-9-19-21-20-18-8-4-5-10-24(18)19;/h2-3,6-7H,4-5,8-15H2,1H3;1H |
| SMILES | Cl.Cc1ccccc1N2CCN(CCc3nnc4CCCCn34)CC2 |
| IUPAC Name | 3-[2-[4-(2-methylphenyl)piperazin-1-yl]ethyl]-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyridine;hydrochloride |
| Molecular Weight (g/mol) | 361.9121 |
| Formula Weight | 361.2033 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction