missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAPP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115093
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DAPP1/BAM32.
May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.
Specifications
| DAPP1 | |
| Polyclonal | |
| Unconjugated | |
| DAPP1 | |
| b lymphocyte adapter protein Bam32; Bam32; B-cell adapter molecule of 32 kDa; Dapp1; Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide; dual adaptor for phosphotyrosine and 3-phosphoinositides 1; dual adaptor of phosphotyrosine and 3-phosphoinositides; dual adaptor of phosphotyrosine and 3-phosphoinositides 1; hDAPP1; HSPC066; mDAPP1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26377, 27071 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9QXT1, Q9UN19 | |
| DAPP1 | |
| A synthesized peptide derived from human DAPP1(Accession Q9UN19), corresponding to amino acid residues P113-S163. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction