missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DARC Antibody (2C3) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-75197-0.025MG
This item is not returnable.
View return policy
Description
DARC Monoclonal specifically detects DARC in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Block/Neutralize.
Specifications
| DARC | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| ACKR1 | |
| CHO cells transfected with cDNA encoding the Duffy glycoprotein. | |
| 2532 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Blocking Assay, Neutralization | |
| 2C3 | |
| Western Blot, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunofluorescence, Block/Neutralize | |
| CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor | |
| Rabbit | |
| 0.025 mg | |
| The antibody binds specifcally to the pentapeptide FEDVW of the Duffy receptor. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction