missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DARPP-32 Antibody (2G6D9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37288-0.025ml
This item is not returnable.
View return policy
Description
DARPP-32 Monoclonal specifically detects DARPP-32 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| DARPP-32 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| protein phosphatase 1, regulatory (inhibitor) subunit 1B, regulatory (inhibitor) subunit 1B (dopamine and cAMPregulated phosphoprotein, DARPP-32) | |
| Mouse | |
| 23 kDa | |
| 0.025 mL | |
| Breast Cancer, Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Phosphatase, Signal Transduction | |
| 84152 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 2G6D9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PPP1R1B | |
| Purified recombinant fragment of human PPP1R1B (AA: 95-204) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction