missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DARS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00267-100ul
This item is not returnable.
View return policy
Description
DARS Polyclonal antibody specifically detects DARS in Human samples. It is validated for ELISA
Specifications
| DARS | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| aspartate tRNA ligase 1, cytoplasmic, Aspartate--tRNA ligase, aspartyl-tRNA synthetase, aspartyl-tRNA synthetase, cytoplasmic, aspRS, Cell proliferation-inducing gene 40 protein, cell proliferation-inducing protein 40, DKFZp781B11202, EC 6.1.1.12, MGC111579 | |
| Produced in rabbits immunized with purified, recombinant Human DARS (Uniprot#: P14868; Met1-Pro501) | |
| 100 μg | |
| DNA replication Transcription Translation and Splicing | |
| 1615 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction