Learn More
Invitrogen™ DARS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527892
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (96%), Rat (94%), Zebrafish (83%), Xenopus laevis (90%), Chicken (90%), Sheep (94%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Aspartyl-tRNA synthetase (DARS) is part of a multienzyme complex of aminoacyl-tRNA synthetases. Aspartyl-tRNA synthetase charges its cognate tRNA with aspartate during protein biosynthesis.
Specifications
| DARS | |
| Polyclonal | |
| Unconjugated | |
| DARS1 | |
| 5730439G15Rik; aspartate tRNA ligase 1, cytoplasmic; aspartate--tRNA ligase, cytoplasmic; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, cytoplasmic; aspRS; Cell proliferation-inducing gene 40 protein; Dars; DRS1; HBSL; PIG40; testicular tissue protein Li 192 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1615 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.35 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P14868 | |
| DARS1 | |
| Recombinant protein encompassing a sequence within the center region of human DARS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.