missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daxx Antibody (PCRP-DAXX-5G11), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14025MFV450
This item is not returnable.
View return policy
Description
Daxx Monoclonal antibody specifically detects Daxx in Human samples. It is validated for Flow Cytometry
Specifications
| Daxx | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-DAXX-5G11 | |
| Flow Cytometry | |
| BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246 | |
| Recombinant full-length human Daxx protein (Uniprot: Q9UER7) | |
| 0.1 mL | |
| Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
| 1616 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction