missing translation for 'onlineSavingsMsg'
Learn More

Zeiss™ Day Light Filter

Use this filter with KL 1500 and KL 2500 LCD Cold Light Source

Brand:  Zeiss™ 000000-1063-317

Additional Details : Weight : 0.50000kg

Product Code. 15324703

  • € 52.00 / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

  • Conversion filter
  • 28mm (D)
Specifications

Specifications

Filter Set D
28 mm
KL1500 and KL2500 LCD
3 grey filters 0.015, 0.06, 0.25
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers