missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAZ2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 462.00
Specifications
| Antigen | DAZ2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DAZ2 Polyclonal antibody specifically detects DAZ2 in Mouse samples. It is validated for Western BlotSpecifications
| DAZ2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| deleted in azoospermia 2, deleted in azoospermia protein 2, MGC126442 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human DAZ2 (NP_001005785.1). MSAANPETPNSTISREASTQSSSAAASQGWVLPEGKIVPNTVFVGGIDARMDETEIGSCFGRYGSVKEVKIITNRTGVSKGYGFVSFVNDVDVQKIVGSQIHFHGKKLKLGPAIRKQKLCARHVQPRPLVVNPPPPPQFQNVWRNPNTETYLQPQITPNPVTQHVQAYSAYPHSPGQVITGCQLLVYNYQEYPTYPDSAF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 57055 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title