missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAZ4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541721
This item is not returnable.
View return policy
Description
Peptide sequence: MSAANPETPN STISREASTQ SSSAAASQGW VLPEGKIVPN TVFVGGIDAR Sequence homology: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 92%; Rat: 100%.
This gene is a member of the DAZ gene family and is a candidate for the human Y-chromosomal azoospermia factor (AZF). This RNA-binding protein is important for spermatogenesis.
Specifications
| DAZ4 | |
| Polyclonal | |
| Unconjugated | |
| DAZ4 | |
| DAZ4; deleted in azoospermia 4; deleted in azoospermia protein 4; pDP1680; pDP1681 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57135 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86SG3 | |
| DAZ4 | |
| Synthetic peptide directed towards the N-terminal of human DAZ4 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction