missing translation for 'onlineSavingsMsg'
Learn More

DBC1 Antibody (8C10) [DyLight 350], Novus Biologicals Biologicals™

Product Code. 30519185 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30519185 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30519185 Supplier Novus Biologicals Supplier No. NBP342956UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DBC1 Monoclonal antibody specifically detects DBC1 in Human,Mouse,Porcine samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DBC1
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 8C10
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias DBCCR1deleted in bladder cancer protein 1, deleted in bladder cancer 1, deleted in bladder cancer chromosome region candidate 1, FAM5AbA574M5.1 (deleted in bladder cancer chromosome region candidate 1 (IB3089A)), Protein FAM5A
Host Species Mouse
Immunogen deleted in bladder cancer 1 - (Uniprot# O60477)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, p53 Pathway, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1620
Target Species Human, Mouse, Pig
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.