missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ DBC1/p30 293T Cell Transient Overexpression Lysate

Product Code. 18425069 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
This item is not returnable. View return policy

Product Code. 18425069

Brand: Novus Biologicals™ H00057805T02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Research Category Cancer, Cell Cycle and Replication, Tumor Suppressors
Target DBC1/p30
Synonym DBC.1, DBC1deleted in breast cancer 1, DBC-1p30 DBC protein, Deleted in breast cancer gene 1 protein, KIAA1967, NET35, p30 DBC, p30DBC
Gene Symbol CCAR2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.