missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DbH Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5119088

 View more versions of this product

Product Code. 17700504

  • € 485.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DbH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene is an oxidoreductase belonging to the copper type II, ascorbate-dependent monooxygenase family. It is present in the synaptic vesicles of postganglionic sympathetic neurons and converts dopamine to norepinephrine. It exists in both soluble and membrane-bound forms, depending on the absence or presence, respectively, of a signal peptide.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DbH
Polyclonal
Unconjugated
DBH
Dbh; DBM; dopamine beta hydroxylase; Dopamine beta hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase precursor; dopamine beta-hydroxylase precursor (EC 1.14.17.1); dopamine beta-monooxygenase; dopamine beta-monooxygenase precursor (EC 1.14.17.1); Dopamine-Beta-hydroxylase; DOPBHY; mixed function oxidase; Soluble dopamine beta-hydroxylase
Rabbit
Antigen affinity chromatography, Protein A
RUO
1621
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P09172
DBH
Recombinant protein Dopamine Beta Hydroxylase. The antigen corresponds to amino acid range 335-571 of the target protein.
200 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.