missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BEX2 Monoclonal Antibody (OTI1D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811154S
This item is not returnable.
View return policy
Description
BEX2 Monoclonal antibody specifically detects BEX2 in Human samples. It is validated for Western BlotSpecifications
| BEX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00213 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 322 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1D1 | |
| Unconjugated | |
| APBB1 | |
| BEX1; BEX2; brain expressed X-linked 2; brain-expressed X-linked protein 2; DJ79P11.1; hBex2; protein BEX2; RP1-79P11.1 | |
| Full length human recombinant protein of human APBB1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction