missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBNL Mouse anti-Human, Clone: OTI7B7, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805346
This item is not returnable.
View return policy
Description
Actin-binding adapter protein. May act as a common effector of antigen receptor-signaling pathways in leukocytes. Its association with dynamin suggests that it may also connect the actin cytoskeleton to endocytic function. Acts as a key component of the immunological synapse that regulates T-cell activation by bridging TCRs and the actin cytoskeleton to gene activation and endocytic processes. Binds to F-actin but is not involved in actin polymerization, capping or bundling. Does not bind G-actin.Specifications
| DBNL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UJU6 | |
| DBNL | |
| Full length human recombit protein of human DBNL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI7B7 | |
| Unconjugated | |
| DBNL | |
| ABP1, HIP-55, HIP55, SH3P7 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 28988 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction