Learn More
DBR1 Mouse anti-Human, CoraLite 488, Clone: 3B5A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66951-100UL
Product Code. 17274716
  • € 450.00 / 100µL
Description
DBR1(Lariat debranching enzyme) hydrolyzes 2-prime-to-5-prime branched phosphodiester bonds at the branch point of excised lariat intron RNA and converts them into linear molecules. DBR1 belongs to the lariat debranching enzyme family. Inhibiton of Dbr1 can suppress TDP-43 toxicity in primary neurons, suggesting that Dbr1 could be a potential therapeutic target for ALS and related TDP-43 proteinopathies (PMID: 23104007). This protein has 2 isoforms produced by alternative splicing.
Specifications
Specifications
DBR1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse
IgG2b
Immunofluorescence
3B5A7
CoraLite 488
100 μL
51163