missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-SIGNR/CD299/CLEC4M Antibody (MM0241-2U25), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12158AF488
This item is not returnable.
View return policy
Description
DC-SIGNR/CD299/CLEC4M Monoclonal specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Flow Cytometry.
Specifications
| DC-SIGNR/CD299/CLEC4M | |
| Monoclonal | |
| Alexa Fluor 488 | |
| NP_001138382 | |
| CLEC4M | |
| Human recombinant DC-SIGNR/CD299/CLEC4M | |
| 0.1 mL | |
| Signal Transduction | |
| 10332 | |
| Store at 4C in the dark. | |
| IgG2 |
| Flow Cytometry, Immunohistochemistry | |
| MM0241-2U25 | |
| Flow Cytometry, Immunohistochemistry | |
| CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction