missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-SIGNR/CD299/CLEC4M Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 574.00
Specifications
| Antigen | DC-SIGNR/CD299/CLEC4M |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667747
|
Novus Biologicals
NBP2-48770-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698675
|
Novus Biologicals
NBP2-48770 |
0.1 mL |
€ 574.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DC-SIGNR/CD299/CLEC4M Polyclonal antibody specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| DC-SIGNR/CD299/CLEC4M | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2), 40% Glycerol | |
| 10332 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GLLEEDPTTSGIRLFPRDFQFQQIHGHKSSTGCLGH | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title