missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC-STAMP/TM7SF4 Antibody (788524), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7824PECY55
This item is not returnable.
View return policy
Description
DC-STAMP/TM7SF4 Monoclonal antibody specifically detects DC-STAMP/TM7SF4 in Human samples. It is validated for Flow Cytometry
Specifications
| DC-STAMP/TM7SF4 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 81501 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry | |
| 788524 | |
| Flow Cytometry | |
| dendritic cell-specific transmembrane protein, transmembrane 7 superfamily member 4 | |
| E. coli-derived recombinant human DC-STAMP, Asp314-Thr376, Accession # Q9H295 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction