missing translation for 'onlineSavingsMsg'
Learn More

DC2 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 204.00 - € 487.00

Specifications

Antigen DC2
Dilution Western Blot 1:100 - 1:500
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18699890
View Documents
Novus Biologicals
NBP2-92350-0.02ml
0.02 mL
€ 204.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18655470
View Documents
Novus Biologicals
NBP2-92350-0.1ml
0.1 mL
€ 487.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DC2 Polyclonal antibody specifically detects DC2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

DC2
Western Blot
Unconjugated
Rabbit
Endocrinology, Signal Transduction
PBS with 50% glycerol, pH7.3.
58505
IgG
Affinity purified
Western Blot 1:100 - 1:500
Polyclonal
Purified
RUO
Human, Mouse, Rat
DC2, Hydrophobic protein HSF-28, oligosaccharyltransferase complex subunit, oligosaccharyltransferase complex subunit OSTC
A synthetic peptide corresponding to a sequence within amino acids 50 to the C-terminus of human OSTC (NP_067050.1). YDVIVEPPSVGSMTDEHGHQRPVAFLAYRVNGQYIMEGLASSFLFTMGGLGFIILDRSNAPNIPKLNRFLLLFIGFVCVLLSFFMARVFMRMKLPGYLMG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.