missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99483-50ul
This item is not returnable.
View return policy
Description
DC8 Polyclonal antibody specifically detects DC8 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| DC8 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:1500-1:50000 | |
| C1orf48, chromosome 1 open reading frame 48, DC8, DKFZP566O1646, kinetochore-associated protein NSL1 homolog, MIS14, NSL1, MIND kinetochore complex component, homolog (S. cerevisiae) | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DC8. | |
| 50 μg | |
| DNA Repair | |
| 25936 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction