missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DC8 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99483PE
This item is not returnable.
View return policy
Description
DC8 Polyclonal antibody specifically detects DC8 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| DC8 | |
| Antigen and protein A Affinity-purified | |
| DNA Repair | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction