missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DCAF12 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17127553
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17127553 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17127553 Supplier Invitrogen™ Supplier No. PA5109619

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MARKVVSRKR KAPASPGAGS DAQGPQFGWD HSLHKRKRLP PVKRSLVYYL KNREVRLQNE TSYSRVLHGY AAQQLPSLLK EREFH.

Cancer-testis (CT) antigens are a type of antigen whose expression is highly restricted, primarily in normal testis, but aberrantly in different tumor types. Their immunogenicity in cancer patients makes them prime targets for immunotherapy of human tumors. One such CT antigen is TCC52, a novel centrosomal protein. TCC52 is a WD repeat-containing protein that interacts with the COP9 signalosome, a macromolecular complex that interacts with cullin-RING E3 ligases and regulates their activity by hydrolyzing cullin-Nedd8 conjugates. The normal biological function of this protein is not known at this time.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCAF12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DCAF12
Gene Accession No. Q5T6F0
Gene Alias 1500001L20Rik; 5830424K06Rik; AA420338; AI851081; cancer/testis antigen 102; centrosome-related protein TCC52; CT102; Dcaf12; DDB1 and CUL4 associated factor 12; DDB1- and CUL4-associated factor 12; Kiaa1892; LOC682812; similar to WD repeat domain 40A; TCC52; Testis cancer centrosome-related protein; WD repeat domain 40A; WD repeat-containing protein 40A; WDR40A
Gene Symbols DCAF12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-85 of human DCAF12 (NP_056212.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25853, 682812
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.