missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DCAF17 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17137553
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17137553 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17137553 Supplier Invitrogen™ Supplier No. PA5109620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LYSFQTIAEQ FMQQKLDLGC ACRWGGTTGT VGEAPFGIPC NIKITDMPPL LFEVSSLENA FQIGGHPWHY IVTPNKKKQK GVFHICALKD NSLAKNGIQE M.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DCAF17
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DCAF17
Gene Accession No. B1H299
Gene Alias 2810055O12Rik; 4833418A01Rik; A030004A10Rik; A930009G19Rik; AI448937; C2orf37; DCAF17; DDB1 and CUL4 associated factor 17; DDB1- and CUL4-associated factor 17; RGD1565551
Gene Symbols DCAF17
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 235-335 of human DCAF17 (NP_079276.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499807
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.