missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC66 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78739AF405
This item is not returnable.
View return policy
Description
CCDC66 Polyclonal antibody specifically detects CCDC66 in Human samples. It is validated for Western Blot
Specifications
| CCDC66 | |
| Polyclonal | |
| Western Blot | |
| coiled-coil domain containing 66, coiled-coil domain-containing protein 66, DKFZp686C0433 | |
| The immunogen for this product maps to a region between residue 898 and 948 of human Coiled-Coil Domain Containing 66 using the numbering given in entry NP_001135419.1 (GeneID 285331). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 285331 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction