missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCAF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141255
This item is not returnable.
View return policy
Description
Suggested positive control: human skeletal muscle protein or tissue.
NRIP is a novel 860a.a nuclear protein consisting of seven conserved WD40 domains and one NLS motif. It binds to androgen and glucocorticoid receptors and up regulates their transcriptional activity, thereby functioning as a nuclear receptor coactivator. Role of NRIP has been implicated in cell growth and also in cervical and prostrate cancer thus indicating a therapeutic intervention. Northern Blot analysis detected a high expression of NRIP in skeletal muscle and testis and low expression in heart, prostrate and adrenal gland.
Specifications
| DCAF6 | |
| Polyclonal | |
| Unconjugated | |
| DCAF6 | |
| 1200006M05Rik; Androgen receptor complex-associated protein; ARCAP; Dcaf6; DDB1 and CUL4 associated factor 6; DDB1- and CUL4-associated factor 6; IQ motif and WD repeat-containing protein 1; IQ motif and WD repeats 1; IQWD1; MSTP055; NRIP; nuclear receptor interaction protein; PC326; RGD1561961 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 289181, 480085, 55827, 74106 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Q58WW2, Q9DC22 | |
| DCAF6 | |
| A portion of amino acids 800-850 of human NRIP. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction