missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCAMKL2 Antibody (2A5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00166614-M01
This item is not returnable.
View return policy
Description
DCAMKL2 Monoclonal antibody specifically detects DCAMKL2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| DCAMKL2 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2A5 | |
| Western Blot 1:500, ELISA | |
| CL2EC 2.7.11.1, CLICK-II, CLIK2, DCDC3, DCDC3BCaMK-like CREB regulatory kinase 2, DCK2DCAMKL2, DKFZp761I032, doublecortin and CaM kinase-like 2, Doublecortin domain-containing protein 3B, Doublecortin-like and CAM kinase-like 2, doublecortin-like kinase 2CLICK2, EC 2.7.11, MGC45428, serine/threonine-protein kinase DCLK2 | |
| DCAMKL2 (AAH32726, 348 a.a. ∽ 438 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. FRGLKISAHGRSSSNVNGGPELDRCISPEGVNGNRCSESSTLLEKYKIGKVIGDGNFAVVKECIDRSTGKEFALKIIDKAKCCGKEHLIE* | |
| 0.1 mg | |
| Signal Transduction | |
| 166614 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction