missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCBLD2/ESDN Antibody (3G10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00131566-M01
This item is not returnable.
View return policy
Description
DCBLD2/ESDN Monoclonal antibody specifically detects DCBLD2/ESDN in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Specifications
| DCBLD2/ESDN | |
| Monoclonal | |
| Unconjugated | |
| NP_563615 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA | |
| 3G10 | |
| In 1x PBS, pH 7.4 | |
| CLCP11700055P21Rik, CUB, LCCL and coagulation factor V/VIII-homology domains protein 1, discoidin, CUB and LCCL domain containing 2, discoidin, CUB and LCCL domain-containing protein 2, Endothelial and smooth muscle cell-derived neuropilin-like protein, ESDNcoagulation factor V/VIII-homology domains protein 1 | |
| DCBLD2 (NP_563615.3, 80 a.a. ∽ 164 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ESGTLTSINYPQTYPNSTVCEWEIRVKMGERVRIKFGDFDIEDSDSCHFNYLRIYNGIGVSRTEIGKYCGLGLQMNHSIESKGNE | |
| 0.1 mg | |
| Signal Transduction | |
| 131566 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction