missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCBLD2/ESDN Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93567V
This item is not returnable.
View return policy
Description
DCBLD2/ESDN Polyclonal antibody specifically detects DCBLD2/ESDN in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DCBLD2/ESDN | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| DyLight 405 | |
| CLCP11700055P21Rik, CUB, LCCL and coagulation factor V/VIII-homology domains protein 1, discoidin, CUB and LCCL domain containing 2, discoidin, CUB and LCCL domain-containing protein 2, Endothelial and smooth muscle cell-derived neuropilin-like protein, ESDNcoagulation factor V/VIII-homology domains protein 1 | |
| Synthetic peptide corresponding to residues 402-419 [QDKIFQGNKDYHKDVRNN] of the human ESDN protein. | |
| 0.1 mL | |
| Signal Transduction | |
| 131566 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction