missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49606-25ul
This item is not returnable.
View return policy
Description
DCC Polyclonal antibody specifically detects DCC in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DCC | |
| Polyclonal | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Colorectal cancer suppressor, CRC18, CRCR1, deleted in colorectal cancer protein, deleted in colorectal carcinoma, IGDCC1colorectal tumor suppressor, Immunoglobulin superfamily DCC subclass member 1, immunoglobulin superfamily, DCC subclass, member 1, netrin receptor DCC, Tumor suppressor protein DCC | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ACVRPTHPLRSFANPLLPPPMSAIEPKVPYTPLLSQPGPTLPKTHVKTASLGLAGKARSPLLPVSVPTAPEVSEESHKPTEDSANVYEQDDLSEQMAS | |
| 25 μL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 1630 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction