missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCIR/CLEC4A Antibody (216110) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1748Z
This item is not returnable.
View return policy
Description
DCIR/CLEC4A Monoclonal antibody specifically detects DCIR/CLEC4A in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| DCIR/CLEC4A | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 216110 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor | |
| Mouse myeloma cell line NS0-derived recombinant human DCIR isoform 1, Gln70-Leu237, Accession # Q9UMR7 | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 50856 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction