missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCIR/CLEC4A Antibody (MM0237-5P20), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12257APC
This item is not returnable.
View return policy
Description
DCIR/CLEC4A Monoclonal specifically detects DCIR/CLEC4A in Human samples. It is validated for Flow Cytometry.
Specifications
| DCIR/CLEC4A | |
| Monoclonal | |
| APC | |
| Q9UMR7 | |
| CLEC4A | |
| Human recombinant DCIR/CLEC4A (extracellular domain) | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 50856 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| MM0237-5P20 | |
| Flow Cytometry | |
| CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction