missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCIR/CLEC4A Antibody (MM0237-5P20), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12257C
This item is not returnable.
View return policy
Description
DCIR/CLEC4A Monoclonal specifically detects DCIR/CLEC4A in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| DCIR/CLEC4A | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CLEC4A | |
| Human recombinant DCIR/CLEC4A (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| MM0237-5P20 | |
| Western Blot, Flow Cytometry | |
| CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor | |
| Mouse | |
| Protein G purified | |
| Immunology, Innate Immunity | |
| 50856 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction