missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCK Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32179AF594
This item is not returnable.
View return policy
Description
DCK Polyclonal antibody specifically detects DCK in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| DCK | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| dCK, deoxycytidine kinase, EC 2.7.1, EC 2.7.1.74, MGC117410, MGC138632 | |
| The immunogen this antibody was made to, maps to a region between residue 210 to 260 of human Deoxycytidine Kinase using the numbering given in entry NP_000779.1 (GeneID 1633). | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 1633 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction