missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCK Monoclonal Antibody (OTI16G6), TrueMAB™, OriGene
Description
DCK Monoclonal antibody specifically detects DCK in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
| Antigen | DCK |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI16G6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | PDHA1 |
| Gene Accession No. | P08559 |
| Gene Alias | DCK; Deoxycytidine kinase; deoxynucleoside kinase |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?