missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCLK1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36420V
This item is not returnable.
View return policy
Description
DCLK1 Polyclonal antibody specifically detects DCLK1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| DCLK1 | |
| Polyclonal | |
| Western Blot | |
| DCAMKL1CLICK1, DCDC3ACL1, DCLK, doublecortin and CaM kinase-like 1, Doublecortin domain-containing protein 3A, doublecortin-like kinase 1Doublecortin-like and CAM kinase-like 1, EC 2.7.11, KIAA0369EC 2.7.11.1, serine/threonine-protein kinase DCLK1 | |
| The immunogen this antibody was made to, maps to a region between residue 690 to 740 of human Doublecortin-Like and CAM Kinase-Like 1 using the numbering given in entry O15075.2 (GeneID 9201). | |
| 0.1 mL | |
| Protein Kinase | |
| 9201 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction