Learn More
DCLK1 Mouse anti-Human, Clone: OTI1F8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF806542
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Doublecortin-like kinase (DCAMKL1, DCLK1)(Ser/Thr protein kinase family) is essential for proper neurogenesis, neuronal migration, and axonal wiring. DCAMKL1 is involved in a calcium-signaling pathway controling neuronal migration in the developing brain, and participates in functions of the mature nervous system. DCAMKL1 protein shares high homology with doublecortin (DCX). DCLK, but not DCX, is highly expressed in regions of active neurogenesis in the neocortex and cerebellum. DCAMKL1 controls mitotic division by regulating spindle formation and also determines the fate of neural progenitors during cortical neurogenesis.Specifications
| DCLK1 | |
| Monoclonal | |
| Unconjugated | |
| DCLK1 | |
| CL1, CLICK1, DCAMKL1, DCDC3A, DCLK | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9201 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1F8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O15075 | |
| DCLK1 | |
| Human recombit protein fragment corresponding to amino acids 193-492 of human DCLK1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.