missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCLRE1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571370
This item is not returnable.
View return policy
Description
DCLRE1A Polyclonal Antibody for Western Blot
May be required for DNA interstrand cross-link repair. Also required for checkpoint mediated cell cycle arrest in early prophase in response to mitotic spindle poisons.
Specifications
| DCLRE1A | |
| Polyclonal | |
| Unconjugated | |
| DCLRE1A | |
| DCLRE1A; DNA cross-link repair 1A; DNA cross-link repair 1A protein; DNA-crosslink repair gene SNM1; hSNM1; hSNM1A; KIAA0086; PSO2; SNM1; SNM1 homolog A; SNM1A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9937 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6PJP8 | |
| DCLRE1A | |
| synthetic peptide directed towards the N terminal region of human DCLRE1A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction