missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCLRE1B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 462.00
Specifications
| Antigen | DCLRE1B |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DCLRE1B Polyclonal antibody specifically detects DCLRE1B in Human, Mouse samples. It is validated for Western BlotSpecifications
| DCLRE1B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| APOLLO, DKFZp686C24187, DNA cross-link repair 1B, DNA cross-link repair 1B (PSO2 homolog, S. cerevisiae), DNA cross-link repair 1B protein, EC 3.1, FLJ12810, FLJ13998, hSNM1B, PSO2 homolog, SNM1 homolog B, SNM1B5' exonuclease Apollo, SNMIB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 323-532 of human DCLRE1B (NP_073747.1). VQQYMSSSSRKPSLLWLLERRLKRPRTQGVVFESPEESADQSQADRDSKKAKKEKLSPWPADLEKQPSHHPLRIKKQLFPDLYSKEWNKAVPFCESQKRVTMLTAPLGFSVHLRSTDEEFISQKTREEIGLGSPLVPMGDDDGGPEATGNQSAWMGHGSPLSHSSKGTPLLATEFRGLALKYLLTPVNFFQAGYSSRRFDQQVEKYHKPC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 64858 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title