missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCLRE1B Antibody (OTI2B7), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72152FR
This item is not returnable.
View return policy
Description
DCLRE1B Monoclonal specifically detects DCLRE1B in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| DCLRE1B | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| DCLRE1B | |
| Recombinant protein fragment corresponding to amino acids 1-280 of human DCLRE1B (NP_073747) produced in E.coli. | |
| 0.1 mL | |
| 64858.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2B7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| APOLLO, DKFZp686C24187, DNA cross-link repair 1B, DNA cross-link repair 1B (PSO2 homolog, S. cerevisiae), DNA cross-link repair 1B protein, EC 3.1, FLJ12810, FLJ13998, hSNM1B, PSO2 homolog, SNM1 homolog B, SNM1B5′ exonuclease Apollo, SNMIB | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction